The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083588.1 | NP_001077057.1 | 2 - 3 | 385 | 169 |
NM_001083589.1 | NP_001077058.1 | 2 - 3 | 236 | 169 | |
NM_001951.3 | NP_001942.2 | 2 - 3 | 385 | 169 | |
GenBank mRNA | AB209185.1 | - | 2 - 3 | 157 | 169 |
AI740797.1 | - | 2 - 3 | 286 | 169 | |
BM919591.1 | - | 2 - 3 | 240 | 169 | |
BP224536.1 | - | 2 - 3 | 236 | 169 | |
U15642.1 | - | 2 - 3 | 551 | 169 | |
U31556.1 | - | 2 - 3 | 386 | 169 | |
X86097.1 | - | 2 - 3 | 378 | 169 | |
Z78409.1 | - | 2 - 3 | 382 | 169 |