Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPC1
Entrez Gene ID
2817
Gene Name
glypican 1
Gene Aliases
glypican
Chromosome Location
Chr.2: 240435698 - 240468078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002081.2 NP_002072.2 6 - 7 1348 91
XM_011510976.2 XP_011509278.1 6 - 7 1249 91
GenBank mRNA AB209122.1 - 4 - 5 3308 91
AK095397.1 - 5 - 6 2244 91
AK096638.1 - 4 - 5 2907 91
BC045712.1 - 6 - 7 1178 91
BC051279.1 - 6 - 7 1348 91
BG825129.1 - 4 - 5 554 91
X54232.1 - 6 - 7 1348 91

View Full Product Details