The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000547.5 | NP_000538.3 | 13 - 14 | 2475 | 70 |
NM_001206744.1 | NP_001193673.1 | 13 - 14 | 2470 | 70 | |
NM_001206745.1 | NP_001193674.1 | 12 - 13 | 2299 | 70 | |
NM_175719.3 | NP_783650.1 | 12 - 13 | 2304 | 70 | |
NM_175722.3 | NP_783653.1 | 11 - 12 | 1866 | 70 | |
XM_011510380.2 | XP_011508682.1 | 12 - 13 | 2385 | 70 | |
GenBank mRNA | AF439430.1 | - | 11 - 12 | 1865 | 70 |
AF533530.1 | - | 4 - 5 | 808 | 70 | |
BC095448.1 | - | 13 - 14 | 2468 | 70 | |
CD608890.1 | - | 1 - 2 | 113 | 70 | |
J02969.1 | - | 13 - 14 | 2456 | 70 | |
J02970.1 | - | 12 - 13 | 2285 | 70 | |
M17755.2 | - | 13 - 14 | 2468 | 70 | |
M55702.1 | - | 2 - 3 | 377 | 70 | |
X17358.1 | - | 12 - 13 | 2278 | 70 | |
Y00406.1 | - | 13 - 14 | 2424 | 70 |