Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCLC
Entrez Gene ID
2729
Gene Name
glutamate-cysteine ligase catalytic subunit
Gene Aliases
GCL, GCS, GLCL, GLCLC
Chromosome Location
Chr.6: 53497341 - 53545129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001498.3 NP_001489.1 4 - 5 1041 97
GenBank mRNA AB262176.1 - 4 - 5 944 97
AK304288.1 - 4 - 5 569 97
AK313480.1 - 4 - 5 951 97
BC022487.1 - 4 - 5 948 97
BC039894.1 - 4 - 5 639 97
M90656.1 - 4 - 5 649 97

View Full Product Details