Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBXN6
Entrez Gene ID
80700
Gene Name
UBX domain protein 6
Gene Aliases
UBXD1, UBXDC2
Chromosome Location
Chr.19: 4445006 - 4458759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171091.1 NP_001164562.1 10 - 11 1199 67
NM_025241.2 NP_079517.1 10 - 11 1301 67
XM_017027325.1 XP_016882814.1 11 - 12 1732 67
GenBank mRNA AF272893.1 - 10 - 11 1300 67
AF272894.1 - 10 - 11 1190 67
AL833695.1 - 3 - 4 1357 67
BC007414.2 - 6 - 7 761 67
BC008288.1 - 10 - 11 1199 67
BC017113.1 - 3 - 4 471 67

View Full Product Details