The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032108.3 | NP_115484.2 | 7 - 8 | 826 | 65 |
XM_011527639.2 | XP_011525941.1 | 7 - 8 | 875 | 65 | |
XM_011527640.2 | XP_011525942.1 | 7 - 8 | 618 | 65 | |
XM_011527641.1 | XP_011525943.1 | 7 - 8 | 875 | 65 | |
GenBank mRNA | AB022433.1 | - | 6 - 7 | 602 | 65 |
AB209278.1 | - | 6 - 7 | 588 | 65 | |
AF216389.1 | - | 6 - 7 | 567 | 65 | |
AF293363.1 | - | 6 - 7 | 567 | 65 | |
AK300037.1 | - | 7 - 8 | 826 | 65 | |
AY358939.1 | - | 6 - 7 | 582 | 65 | |
BC142617.1 | - | 7 - 8 | 769 | 65 | |
FR839673.1 | - | 6 - 7 | 564 | 65 |