Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA6B
Entrez Gene ID
10501
Gene Name
semaphorin 6B
Gene Aliases
SEM-SEMA-Y, SEMA-VIB, SEMAN, semaZ
Chromosome Location
Chr.19: 4542588 - 4581484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032108.3 NP_115484.2 10 - 11 1249 69
XM_011527639.2 XP_011525941.1 10 - 11 1298 69
XM_011527640.2 XP_011525942.1 10 - 11 1041 69
XM_011527641.1 XP_011525943.1 10 - 11 1298 69
GenBank mRNA AB022433.1 - 9 - 10 1025 69
AB209278.1 - 9 - 10 1011 69
AF216389.1 - 9 - 10 990 69
AF293363.1 - 9 - 10 990 69
AK300037.1 - 10 - 11 1249 69
AY358939.1 - 9 - 10 1005 69
BC142617.1 - 10 - 11 1192 69
FR839673.1 - 9 - 10 987 69

View Full Product Details