The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002841.3 | NP_002832.3 | 12 - 13 | 2872 | 113 |
XM_005265353.3 | XP_005265410.1 | 12 - 13 | 2530 | 113 | |
XM_017006961.1 | XP_016862450.1 | 13 - 14 | 2388 | 113 | |
XM_017006962.1 | XP_016862451.1 | 13 - 14 | 2207 | 113 | |
XM_017006963.1 | XP_016862452.1 | 13 - 14 | 2388 | 113 | |
XM_017006964.1 | XP_016862453.1 | 12 - 13 | 3258 | 113 | |
GenBank mRNA | AB209871.1 | - | 12 - 13 | 2518 | 113 |
BC140904.1 | - | 12 - 13 | 2530 | 113 | |
BC144110.1 | - | 12 - 13 | 2530 | 113 | |
EU273472.1 | - | 1 - 2 | 569 | 113 | |
L09247.1 | - | 12 - 13 | 2232 | 113 |