The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002841.3 | NP_002832.3 | 13 - 14 | 3004 | 75 |
XM_017006961.1 | XP_016862450.1 | 14 - 15 | 2520 | 75 | |
XM_017006962.1 | XP_016862451.1 | 14 - 15 | 2339 | 75 | |
XM_017006964.1 | XP_016862453.1 | 13 - 14 | 3390 | 75 | |
GenBank mRNA | AB209871.1 | - | 13 - 14 | 2650 | 75 |
BC140904.1 | - | 13 - 14 | 2662 | 75 | |
L09247.1 | - | 13 - 14 | 2364 | 75 |