Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NECAP2
Entrez Gene ID
55707
Gene Name
NECAP endocytosis associated 2
Gene Aliases
-
Chromosome Location
Chr.1: 16440672 - 16460090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145278.1 NP_001138750.1 6 - 7 682 54
NM_018090.4 NP_060560.1 6 - 7 760 54
GenBank mRNA AK001282.1 - 6 - 7 692 54
AK021938.1 - 2 - 3 236 54
AK024468.1 - 5 - 6 3312 54
AK302227.1 - 6 - 7 628 54
BC017014.1 - 6 - 7 691 54
BC018914.2 - 6 - 7 694 54

View Full Product Details