Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRF
Entrez Gene ID
5792
Gene Name
protein tyrosine phosphatase, receptor type F
Gene Aliases
BNAH2, LAR
Chromosome Location
Chr.1: 43522238 - 43623672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005271079.3 XP_005271136.1 27 - 28 4771 70
XM_005271080.3 XP_005271137.1 26 - 27 4759 70
XM_005271081.3 XP_005271138.1 25 - 26 4732 70
XM_005271082.3 XP_005271139.1 23 - 24 4393 70
XM_006710795.3 XP_006710858.1 28 - 29 4749 70
XM_006710796.3 XP_006710859.1 26 - 27 4744 70
XM_006710797.3 XP_006710860.1 26 - 27 4738 70
XM_006710798.3 XP_006710861.1 25 - 26 4711 70
XM_006710799.3 XP_006710862.1 26 - 27 4465 70
XM_006710800.3 XP_006710863.1 25 - 26 4438 70
XM_006710801.3 XP_006710864.1 26 - 27 4192 70
XM_011541871.2 XP_011540173.1 28 - 29 4783 70
XM_011541872.2 XP_011540174.1 28 - 29 4769 70
XM_011541873.2 XP_011540175.1 28 - 29 4703 70
XM_017001942.1 XP_016857431.1 25 - 26 4400 70
XM_017001943.1 XP_016857432.1 25 - 26 4453 70
XM_017001944.1 XP_016857433.1 24 - 25 4405 70
XM_017001945.1 XP_016857434.1 24 - 25 4426 70
XM_017001946.1 XP_016857435.1 25 - 26 3886 70
XM_017001947.1 XP_016857436.1 24 - 25 3859 70
XM_017001948.1 XP_016857437.1 24 - 25 3885 70
XM_017001949.1 XP_016857438.1 25 - 26 3850 70
XM_017001950.1 XP_016857439.1 22 - 23 3822 70
GenBank mRNA AB177856.1 - 25 - 26 3383 70
AB177857.1 - 24 - 25 4658 70
AB209478.1 - 16 - 17 3307 70
AK125325.1 - 16 - 17 2643 70
AK127007.1 - 13 - 14 1586 70
AK127475.1 - 3 - 4 875 70
BC048768.1 - 24 - 25 4669 70
BX537361.1 - 12 - 13 2870 70
Y00815.1 - 25 - 26 4710 70

View Full Product Details