The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003660.3 | NP_003651.1 | 6 - 7 | 1010 | 56 |
NR_103842.1 | 6 - 7 | 1010 | 56 | ||
XM_017027407.1 | XP_016882896.1 | 6 - 7 | 1010 | 56 | |
GenBank mRNA | AB014554.2 | - | 6 - 7 | 993 | 56 |
AK025972.1 | - | 3 - 4 | 210 | 56 | |
AK289757.1 | - | 6 - 7 | 715 | 56 | |
AK293798.1 | - | 4 - 5 | 461 | 56 | |
AK308483.1 | - | 4 - 5 | 501 | 56 | |
BC101518.1 | - | 6 - 7 | 736 | 56 | |
BC101520.1 | - | 6 - 7 | 736 | 56 | |
BC143470.1 | - | 6 - 7 | 736 | 56 |