The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081675.2 | NP_001075144.2 | 2 - 3 | 1484 | 82 |
XM_005250901.4 | XP_005250958.1 | 3 - 4 | 1705 | 82 | |
GenBank mRNA | AB196636.1 | - | 2 - 3 | 1460 | 82 |
BC127886.1 | - | 2 - 3 | 1484 | 82 |