The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136493.2 | NP_001129965.1 | 4 - 5 | 697 | 66 |
NM_001287808.1 | NP_001274737.1 | 2 - 3 | 398 | 66 | |
NM_001287809.1 | NP_001274738.1 | 4 - 5 | 547 | 66 | |
NM_032793.4 | NP_116182.2 | 4 - 5 | 658 | 66 | |
NR_109896.1 | 4 - 5 | 658 | 66 | ||
XM_005271285.1 | XP_005271342.1 | 4 - 5 | 622 | 66 | |
XM_011542312.2 | XP_011540614.1 | 4 - 5 | 625 | 66 | |
GenBank mRNA | AB073383.1 | - | 4 - 5 | 622 | 66 |
AF289609.1 | - | 4 - 5 | 614 | 66 | |
AF370364.1 | - | 1 - 2 | 1123 | 66 | |
AK074186.1 | - | 4 - 5 | 563 | 66 | |
AK075183.1 | - | 4 - 5 | 625 | 66 | |
AK093223.1 | - | 4 - 5 | 659 | 66 | |
AK291540.1 | - | 4 - 5 | 620 | 66 | |
AK297991.1 | - | 4 - 5 | 511 | 66 | |
AK301533.1 | - | 2 - 3 | 365 | 66 | |
AL834184.1 | - | 4 - 5 | 612 | 66 | |
AY358636.1 | - | 4 - 5 | 639 | 66 | |
BC011587.1 | - | 4 - 5 | 632 | 66 | |
BC092414.1 | - | 4 - 5 | 600 | 66 |