Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SETD5
Entrez Gene ID
55209
Gene Name
SET domain containing 5
Gene Aliases
-
Chromosome Location
Chr.3: 9397700 - 9478154 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080517.2 NP_001073986.1 15 - 16 2548 74
NM_001292043.1 NP_001278972.1 17 - 18 2812 74
XM_005265301.1 XP_005265358.1 15 - 16 2586 74
XM_005265303.1 XP_005265360.1 17 - 18 2967 74
XM_011533920.1 XP_011532222.1 18 - 19 3062 74
XM_011533921.1 XP_011532223.1 17 - 18 2664 74
XM_011533922.1 XP_011532224.1 16 - 17 2625 74
XM_011533927.1 XP_011532229.1 18 - 19 3005 74
XM_011533928.1 XP_011532230.1 16 - 17 2568 74
XM_011533930.1 XP_011532232.1 18 - 19 3006 74
XM_011533932.1 XP_011532234.1 16 - 17 2773 74
XM_011533933.1 XP_011532235.1 14 - 15 2142 74
XM_017006767.1 XP_016862256.1 18 - 19 4724 74
XM_017006768.1 XP_016862257.1 16 - 17 4238 74
XM_017006769.1 XP_016862258.1 16 - 17 6341 74
XM_017006770.1 XP_016862259.1 17 - 18 6380 74
XM_017006771.1 XP_016862260.1 15 - 16 2586 74
XM_017006772.1 XP_016862261.1 16 - 17 6341 74
XM_017006773.1 XP_016862262.1 16 - 17 6284 74
XM_017006774.1 XP_016862263.1 15 - 16 2529 74
XM_017006775.1 XP_016862264.1 16 - 17 6284 74
XM_017006776.1 XP_016862265.1 17 - 18 2869 74
XM_017006777.1 XP_016862266.1 17 - 18 2812 74
XM_017006778.1 XP_016862267.1 18 - 19 2853 74
XM_017006779.1 XP_016862268.1 16 - 17 2830 74
XM_017006780.1 XP_016862269.1 17 - 18 2814 74
XM_017006781.1 XP_016862270.1 18 - 19 2853 74
XM_017006782.1 XP_016862271.1 17 - 18 6380 74
XM_017006783.1 XP_016862272.1 19 - 20 3078 74
XM_017006784.1 XP_016862273.1 17 - 18 6380 74
XM_017006785.1 XP_016862274.1 17 - 18 6380 74
XM_017006786.1 XP_016862275.1 17 - 18 6380 74
GenBank mRNA AB051544.1 - 11 - 12 2342 74
AK001569.1 - 4 - 5 731 74
AK024384.1 - 3 - 4 642 74
AK091393.1 - 12 - 13 1851 74
AK127298.1 - 5 - 6 1435 74
AK289529.1 - 6 - 7 1130 74
AL442073.1 - 2 - 3 528 74
BC020956.1 - 4 - 5 762 74
BX648380.1 - 7 - 8 1150 74
CR627408.1 - 14 - 15 2164 74

View Full Product Details