Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL5A2
Entrez Gene ID
1290
Gene Name
collagen type V alpha 2 chain
Gene Aliases
EDSC
Chromosome Location
Chr.2: 189031915 - 189225301 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000393.3 NP_000384.2 39 - 40 2937 67
XM_011510573.2 XP_011508875.1 39 - 40 2719 67
GenBank mRNA AB209045.1 - 39 - 40 2799 67
BC086874.1 - 23 - 24 1926 67
X04758.1 - 21 - 22 1462 67
Y14690.1 - 39 - 40 2791 67

View Full Product Details