Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL5A2
Entrez Gene ID
1290
Gene Name
collagen type V alpha 2 chain
Gene Aliases
EDSC
Chromosome Location
Chr.2: 189031915 - 189225301 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000393.3 NP_000384.2 9 - 10 966 76
XM_011510573.2 XP_011508875.1 9 - 10 748 76
GenBank mRNA AB209045.1 - 9 - 10 828 76
AK297936.1 - 9 - 10 848 76
J04478.1 - 9 - 10 829 76
Y14690.1 - 9 - 10 829 76

View Full Product Details