The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042368.2 | NP_001035827.1 | 15 - 16 | 2289 | 93 |
NM_001271774.1 | NP_001258703.1 | 15 - 16 | 2201 | 93 | |
NM_001271775.1 | NP_001258704.1 | 15 - 16 | 2294 | 93 | |
NM_001271776.1 | NP_001258705.1 | 15 - 16 | 2274 | 93 | |
NM_006266.3 | NP_006257.1 | 15 - 16 | 2310 | 93 | |
GenBank mRNA | AF027169.1 | - | 15 - 16 | 2205 | 93 |
AF295773.1 | - | 15 - 16 | 2225 | 93 | |
AK000242.1 | - | 10 - 11 | 1267 | 93 | |
AK024575.1 | - | 7 - 8 | 864 | 93 | |
AK074114.1 | - | 15 - 16 | 2180 | 93 | |
AK090450.1 | - | 11 - 12 | 1636 | 93 | |
AK127524.1 | - | 15 - 16 | 2294 | 93 | |
AK301470.1 | - | 15 - 16 | 2161 | 93 | |
BC021581.1 | - | 3 - 4 | 386 | 93 | |
BC033198.1 | - | 7 - 8 | 706 | 93 | |
BC059362.1 | - | 15 - 16 | 2264 | 93 | |
U14417.1 | - | 4 - 5 | 456 | 93 |