Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR5A2
Entrez Gene ID
2494
Gene Name
nuclear receptor subfamily 5 group A member 2
Gene Aliases
B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2
Chromosome Location
Chr.1: 200027602 - 200177424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205860.2 NP_995582.1 1 - 2 314 106
GenBank mRNA AB019246.1 - 1 - 2 194 106
AF124247.1 - 1 - 2 68 106
U93553.1 - 1 - 2 311 105

View Full Product Details