Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC37
Entrez Gene ID
9652
Gene Name
tetratricopeptide repeat domain 37
Gene Aliases
KIAA0372, Ski3, THES
Chromosome Location
Chr.5: 95463895 - 95555005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014639.3 NP_055454.1 33 - 34 3858 84
GenBank mRNA AB002370.1 - 33 - 34 3858 84
BC056893.1 - 33 - 34 3831 84

View Full Product Details