Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUNDC3A
Entrez Gene ID
10900
Gene Name
RUN domain containing 3A
Gene Aliases
RAP2IP, RPIP-8, RPIP8
Chromosome Location
Chr.17: 44308559 - 44318670 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144825.1 NP_001138297.1 7 - 8 1073 88
NM_001144826.1 NP_001138298.1 7 - 8 1058 88
NM_006695.4 NP_006686.1 7 - 8 1073 88
XM_005256964.3 XP_005257021.1 7 - 8 1006 88
XM_017024035.1 XP_016879524.1 7 - 8 1073 88
XM_017024036.1 XP_016879525.1 7 - 8 994 88
XM_017024037.1 XP_016879526.1 7 - 8 1006 88
XM_017024038.1 XP_016879527.1 7 - 8 1073 88
XM_017024039.1 XP_016879528.1 7 - 8 1073 88
XM_017024040.1 XP_016879529.1 7 - 8 994 88
GenBank mRNA AB209802.1 - 7 - 8 1057 88
AF055026.1 - 7 - 8 837 88
AK123330.1 - 6 - 7 2718 88
AK226045.1 - 7 - 8 1034 88
AK313669.1 - 7 - 8 994 88
AL133657.1 - 7 - 8 1219 88
BC006194.2 - 7 - 8 1036 88
BX537443.1 - 7 - 8 955 88
CR456748.1 - 7 - 8 799 88
U93871.1 - 7 - 8 784 88

View Full Product Details