The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021213.3 | NP_067036.2 | 1 - 2 | 265 | 80 |
XM_005257549.1 | XP_005257606.1 | 1 - 2 | 265 | 80 | |
XM_005257550.3 | XP_005257607.1 | 1 - 2 | 265 | 80 | |
GenBank mRNA | AF087857.1 | - | 1 - 2 | 250 | 80 |
AF114430.1 | - | 1 - 2 | 155 | 80 | |
AF151638.1 | - | 1 - 2 | 249 | 80 | |
AK024667.1 | - | 1 - 2 | 191 | 80 | |
AK314509.1 | - | 1 - 2 | 191 | 80 | |
BC012084.1 | - | 1 - 2 | 183 | 80 | |
BM153142.1 | - | 1 - 2 | 265 | 80 |