The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005257550.3 | XP_005257607.1 | 5 - 6 | 698 | 80 |
XM_017024913.1 | XP_016880402.1 | 5 - 6 | 780 | 80 |