The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136554.1 | NP_001130026.1 | 17 - 18 | 1801 | 135 |
NM_001136555.1 | NP_001130027.1 | 15 - 16 | 1572 | 135 | |
NM_012290.4 | NP_036422.3 | 17 - 18 | 2242 | 135 | |
XM_011512238.2 | XP_011510540.1 | 16 - 17 | 2093 | 135 | |
XM_017005421.1 | XP_016860910.1 | 18 - 19 | 2351 | 135 | |
XM_017005422.1 | XP_016860911.1 | 16 - 17 | 2163 | 135 | |
GenBank mRNA | AB004885.1 | - | 18 - 19 | 2045 | 135 |
AF162666.1 | - | 17 - 18 | 1923 | 135 | |
AF246219.1 | - | 10 - 11 | 1128 | 135 | |
AK090779.1 | - | 17 - 18 | 1801 | 135 | |
AK091975.1 | - | 18 - 19 | 2500 | 135 | |
AK292508.1 | - | 17 - 18 | 1883 | 135 | |
AK301857.1 | - | 15 - 16 | 1572 | 135 | |
BC032657.1 | - | 17 - 18 | 1803 | 135 | |
D50927.2 | - | 17 - 18 | 1875 | 135 | |
Z25421.1 | - | 1 - 2 | 168 | 135 |