Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC11A2
Entrez Gene ID
4891
Gene Name
solute carrier family 11 member 2
Gene Aliases
AHMIO1, DCT1, DMT1, NRAMP2
Chromosome Location
Chr.12: 50952263 - 51028335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000617.2 NP_000608.1 8 - 9 825 80
NM_001174125.1 NP_001167596.1 8 - 9 817 80
NM_001174126.1 NP_001167597.1 8 - 9 825 80
NM_001174127.1 NP_001167598.1 8 - 9 820 80
NM_001174128.1 NP_001167599.1 8 - 9 820 80
NM_001174129.1 NP_001167600.1 8 - 9 1085 80
NM_001174130.1 NP_001167601.1 7 - 8 800 80
NR_033421.1 8 - 9 745 80
NR_033422.1 8 - 9 820 80
XM_005268911.3 XP_005268968.1 8 - 9 877 80
XM_005268912.4 XP_005268969.1 7 - 8 1002 80
XM_011538404.2 XP_011536706.1 8 - 9 1201 80
XM_011538405.2 XP_011536707.1 8 - 9 809 80
XM_017019355.1 XP_016874844.1 8 - 9 1085 80
XM_017019356.1 XP_016874845.1 8 - 9 1015 80
GenBank mRNA AB004857.1 - 8 - 9 768 80
AB062284.1 - 8 - 9 742 80
AF046997.1 - 8 - 9 778 80
AF064484.1 - 8 - 9 768 80
AJ493662.1 - 8 - 9 817 80
AK094735.1 - 8 - 9 1085 80
AK128640.1 - 8 - 9 717 80
AK296445.1 - 7 - 8 779 80
AK315987.1 - 8 - 9 731 80
AK316507.1 - 7 - 8 756 80
BC002592.2 - 8 - 9 772 80
BC100014.1 - 8 - 9 793 80
CK000292.1 - 4 - 5 276 80
KT455015.1 - 8 - 9 781 80
L37347.1 - 5 - 6 518 80

View Full Product Details