Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CWF19L2
Entrez Gene ID
143884
Gene Name
CWF19-like 2, cell cycle control (S. pombe)
Gene Aliases
-
Chromosome Location
Chr.11: 107326345 - 107457846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152434.2 NP_689647.2 8 - 9 1465 98
GenBank mRNA AK056905.1 - 6 - 7 1117 98
AK126881.1 - 8 - 9 1457 98
AK293001.1 - 8 - 9 1400 98
BC110439.1 - 5 - 6 1078 98
BC110440.1 - 5 - 6 1078 98
BC110441.1 - 5 - 6 1078 98
BC110442.1 - 5 - 6 1078 98
BC118669.1 - 5 - 6 1081 98
BX647832.1 - 8 - 9 1439 98
BX648704.1 - 8 - 9 1444 98
BX649022.1 - 8 - 9 1435 98

View Full Product Details