The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144829.2 | NP_001138301.1 | 3 - 4 | 725 | 139 |
NM_001144830.2 | NP_001138302.1 | 2 - 3 | 935 | 139 | |
NM_001461.3 | NP_001452.2 | 3 - 4 | 715 | 139 | |
XM_005272946.4 | XP_005273003.1 | 3 - 4 | 689 | 139 | |
XM_005272947.4 | XP_005273004.1 | 3 - 4 | 433 | 139 | |
XM_005272948.4 | XP_005273005.1 | 3 - 4 | 416 | 139 | |
XM_006711244.3 | XP_006711307.1 | 3 - 4 | 705 | 139 | |
XM_006711245.3 | XP_006711308.1 | 3 - 4 | 705 | 139 | |
XM_011509350.2 | XP_011507652.1 | 3 - 4 | 720 | 139 | |
GenBank mRNA | AK022172.1 | - | 2 - 3 | 925 | 139 |
AK222728.1 | - | 3 - 4 | 430 | 139 | |
AK225739.1 | - | 3 - 4 | 440 | 139 | |
AK314647.1 | - | 3 - 4 | 643 | 139 | |
BC035687.1 | - | 2 - 3 | 608 | 139 | |
L37080.1 | - | 3 - 4 | 401 | 139 | |
Z47553.1 | - | 3 - 4 | 407 | 139 |