Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOA2
Entrez Gene ID
10499
Gene Name
nuclear receptor coactivator 2
Gene Aliases
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
Chromosome Location
Chr.8: 70109762 - 70405390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321703.1 NP_001308632.1 16 - 17 3615 75
NM_001321707.1 NP_001308636.1 16 - 17 3461 75
NM_001321711.1 NP_001308640.1 15 - 16 3336 75
NM_001321712.1 NP_001308641.1 15 - 16 3253 75
NM_001321713.1 NP_001308642.1 15 - 16 3356 75
NM_006540.3 NP_006531.1 16 - 17 3543 75
XM_017012961.1 XP_016868450.1 16 - 17 3530 75
XM_017012962.1 XP_016868451.1 16 - 17 3554 75
XM_017012963.1 XP_016868452.1 16 - 17 4767 75
XM_017012964.1 XP_016868453.1 16 - 17 3822 75
XM_017012965.1 XP_016868454.1 16 - 17 3487 75
XM_017012966.1 XP_016868455.1 17 - 18 3751 75
XM_017012967.1 XP_016868456.1 15 - 16 3465 75
XM_017012968.1 XP_016868457.1 17 - 18 3552 75
XM_017012969.1 XP_016868458.1 15 - 16 3583 75
XM_017012970.1 XP_016868459.1 15 - 16 10002 75
XM_017012971.1 XP_016868460.1 15 - 16 3714 75
XM_017012972.1 XP_016868461.1 12 - 13 3045 75
GenBank mRNA AK298528.1 - 4 - 5 628 75
BC114383.1 - 15 - 16 3410 75
X97674.1 - 16 - 17 3488 75

View Full Product Details