Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOA2
Entrez Gene ID
10499
Gene Name
nuclear receptor coactivator 2
Gene Aliases
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
Chromosome Location
Chr.8: 70109762 - 70405390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321703.1 NP_001308632.1 21 - 22 4579 73
NM_001321707.1 NP_001308636.1 21 - 22 4425 73
NM_001321711.1 NP_001308640.1 20 - 21 4300 73
NM_001321712.1 NP_001308641.1 20 - 21 4217 73
NM_001321713.1 NP_001308642.1 20 - 21 4320 73
NM_006540.3 NP_006531.1 21 - 22 4507 73
XM_017012961.1 XP_016868450.1 21 - 22 4701 73
XM_017012962.1 XP_016868451.1 21 - 22 4725 73
XM_017012963.1 XP_016868452.1 21 - 22 5938 73
XM_017012964.1 XP_016868453.1 21 - 22 4993 73
XM_017012965.1 XP_016868454.1 21 - 22 4658 73
XM_017012966.1 XP_016868455.1 22 - 23 4922 73
XM_017012967.1 XP_016868456.1 20 - 21 4636 73
XM_017012968.1 XP_016868457.1 22 - 23 4723 73
XM_017012969.1 XP_016868458.1 20 - 21 4754 73
XM_017012970.1 XP_016868459.1 20 - 21 11173 73
XM_017012971.1 XP_016868460.1 20 - 21 4885 73
XM_017012972.1 XP_016868461.1 17 - 18 4216 73
GenBank mRNA AK298528.1 - 9 - 10 1592 73
BC114383.1 - 20 - 21 4374 73
X97674.1 - 21 - 22 4452 73

View Full Product Details