The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018672.4 | NP_061142.2 | 12 - 13 | 3002 | 140 |
NM_172232.3 | NP_758424.1 | 13 - 14 | 1930 | 140 | |
GenBank mRNA | AJ275973.3 | - | 13 - 14 | 1849 | 140 |
AJ512612.1 | - | 13 - 14 | 1847 | 140 | |
AK292592.1 | - | 14 - 15 | 2049 | 140 | |
AY028897.1 | - | 12 - 13 | 2794 | 140 | |
BX640746.1 | - | 6 - 7 | 797 | 140 |