Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DCTN1
- Entrez Gene ID
-
1639
- Gene Name
- dynactin subunit 1
- Gene Aliases
-
DAP-150, DP-150, P135
- Chromosome Location
-
Chr.2: 74361154 - 74392087 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U73799.1 |
- |
1 - 2 |
301 |
64 |
View Full Product Details