Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TET3
Entrez Gene ID
200424
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
hCG_40738
Chromosome Location
Chr.2: 73982036 - 74108176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287491.1 NP_001274420.1 3 - 4 2495 122
XM_005264187.3 XP_005264244.1 4 - 5 3015 122
XM_011532682.2 XP_011530984.1 4 - 5 3015 122
XM_011532683.2 XP_011530985.1 4 - 5 3324 122
XM_011532684.2 XP_011530986.1 4 - 5 4756 122
XM_011532685.2 XP_011530987.1 3 - 4 2958 122
XM_011532686.2 XP_011530988.1 3 - 4 2958 122
XM_011532687.2 XP_011530989.1 3 - 4 2297 122
XM_011532688.2 XP_011530990.1 4 - 5 3015 122
XM_017003566.1 XP_016859055.1 4 - 5 4855 122
XM_017003567.1 XP_016859056.1 3 - 4 2327 122
GenBank mRNA AF466365.1 - 1 - 2 1253 122
HQ220209.1 - 3 - 4 2495 122

View Full Product Details