Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNKS
Entrez Gene ID
8658
Gene Name
tankyrase
Gene Aliases
ARTD5, PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5
Chromosome Location
Chr.8: 9555229 - 9782346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003747.2 NP_003738.2 17 - 18 2642 101
XM_006716263.3 XP_006716326.1 17 - 18 3348 101
XM_011543845.2 XP_011542147.1 17 - 18 3348 101
XM_011543846.2 XP_011542148.1 17 - 18 3348 101
XM_011543847.2 XP_011542149.1 15 - 16 3035 101
GenBank mRNA AB209339.1 - 16 - 17 1822 101
AF082556.1 - 17 - 18 2642 101
AF082557.1 - 16 - 17 1938 101
AK302534.1 - 17 - 18 2316 101
BC098394.1 - 17 - 18 2665 101

View Full Product Details