Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SETD5
Entrez Gene ID
55209
Gene Name
SET domain containing 5
Gene Aliases
-
Chromosome Location
Chr.3: 9397700 - 9478154 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080517.2 NP_001073986.1 2 - 3 341 104
NM_001292043.1 NP_001278972.1 2 - 3 341 104
XM_005265301.1 XP_005265358.1 2 - 3 322 104
XM_005265303.1 XP_005265360.1 2 - 3 341 104
XM_011533920.1 XP_011532222.1 2 - 3 341 104
XM_011533921.1 XP_011532223.1 2 - 3 168 104
XM_011533922.1 XP_011532224.1 2 - 3 322 104
XM_011533927.1 XP_011532229.1 2 - 3 341 104
XM_011533928.1 XP_011532230.1 2 - 3 322 104
XM_011533930.1 XP_011532232.1 2 - 3 341 104
XM_011533932.1 XP_011532234.1 2 - 3 341 104
XM_017006767.1 XP_016862256.1 2 - 3 341 104
XM_017006769.1 XP_016862258.1 2 - 3 341 104
XM_017006770.1 XP_016862259.1 2 - 3 341 104
XM_017006771.1 XP_016862260.1 2 - 3 322 104
XM_017006772.1 XP_016862261.1 2 - 3 341 104
XM_017006773.1 XP_016862262.1 2 - 3 341 104
XM_017006774.1 XP_016862263.1 2 - 3 322 104
XM_017006775.1 XP_016862264.1 2 - 3 341 104
XM_017006776.1 XP_016862265.1 2 - 3 341 104
XM_017006777.1 XP_016862266.1 2 - 3 341 104
XM_017006778.1 XP_016862267.1 2 - 3 341 104
XM_017006779.1 XP_016862268.1 2 - 3 341 104
XM_017006780.1 XP_016862269.1 2 - 3 341 104
XM_017006781.1 XP_016862270.1 2 - 3 341 104
XM_017006782.1 XP_016862271.1 2 - 3 341 104
XM_017006783.1 XP_016862272.1 2 - 3 341 104
XM_017006784.1 XP_016862273.1 2 - 3 341 104
XM_017006785.1 XP_016862274.1 2 - 3 341 104
XM_017006786.1 XP_016862275.1 2 - 3 341 104
GenBank mRNA AK092046.1 - 2 - 3 141 104
DB275141.1 - 2 - 3 343 104

View Full Product Details