Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEG
Entrez Gene ID
10290
Gene Name
SPEG complex locus
Gene Aliases
APEG-1, APEG1, BPEG, CNM5, SPEGalpha, SPEGbeta
Chromosome Location
Chr.2: 219434215 - 219493632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005876.4 NP_005867.3 5 - 6 2254 126
XM_005246237.2 XP_005246294.1 7 - 8 1982 126
XM_005246239.2 XP_005246296.1 7 - 8 1797 126
XM_006712189.3 XP_006712252.1 6 - 7 1987 126
XM_011510479.2 XP_011508781.1 6 - 7 2420 126
XM_011510483.2 XP_011508785.2 6 - 7 2017 126
XM_017003157.1 XP_016858646.1 6 - 7 2017 126
XM_017003159.1 XP_016858648.1 3 - 4 1776 126
XM_017003161.1 XP_016858650.1 3 - 4 1776 126
XM_017003162.1 XP_016858651.1 3 - 4 1776 126
GenBank mRNA AK055387.1 - 1 - 2 607 126
AK126500.1 - 6 - 7 2442 126
AK128689.1 - 5 - 6 2252 126
AK289531.1 - 7 - 8 1779 126
AY603755.1 - 4 - 5 1867 126

View Full Product Details