The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173476.1 | NP_001166947.1 | 1 - 2 | 187 | 106 |
XM_005246240.2 | XP_005246297.1 | 1 - 2 | 187 | 106 | |
GenBank mRNA | DA063220.1 | - | 1 - 2 | 188 | 106 |