The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000751.2 | NP_000742.1 | 2 - 3 | 253 | 77 |
NM_001311195.1 | NP_001298124.1 | 2 - 3 | 253 | 77 | |
NM_001311196.1 | NP_001298125.1 | 2 - 3 | 253 | 77 | |
XM_011510524.2 | XP_011508826.1 | 2 - 3 | 225 | 77 | |
GenBank mRNA | AK291526.1 | - | 2 - 3 | 225 | 77 |
AK296704.1 | - | 2 - 3 | 225 | 77 | |
AK304890.1 | - | 2 - 3 | 225 | 77 | |
AK315297.1 | - | 2 - 3 | 225 | 77 | |
BC093923.1 | - | 2 - 3 | 229 | 77 | |
BC093925.1 | - | 2 - 3 | 229 | 77 | |
CD013895.1 | - | 2 - 3 | 198 | 77 | |
X55019.1 | - | 2 - 3 | 201 | 77 |