The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015338.5 | NP_056153.2 | 9 - 10 | 1319 | 60 |
XM_006723727.3 | XP_006723790.1 | 8 - 9 | 1598 | 60 | |
XM_006723728.3 | XP_006723791.1 | 9 - 10 | 928 | 60 | |
XM_006723730.3 | XP_006723793.1 | 9 - 10 | 1124 | 60 | |
XM_006723732.2 | XP_006723795.1 | 8 - 9 | 947 | 60 | |
XM_006723733.1 | XP_006723796.1 | 2 - 3 | 953 | 60 | |
XM_011528648.2 | XP_011526950.1 | 9 - 10 | 1243 | 60 | |
XM_011528652.2 | XP_011526954.1 | 10 - 11 | 2261 | 60 | |
XM_017027704.1 | XP_016883193.1 | 9 - 10 | 2384 | 60 | |
XM_017027705.1 | XP_016883194.1 | 9 - 10 | 1397 | 60 | |
XM_017027706.1 | XP_016883195.1 | 8 - 9 | 1448 | 60 | |
GenBank mRNA | AB023195.2 | - | 3 - 4 | 369 | 60 |
AB084280.1 | - | 8 - 9 | 872 | 60 | |
AJ438952.2 | - | 9 - 10 | 1145 | 60 | |
AK122923.1 | - | 8 - 9 | 947 | 60 | |
AL117518.1 | - | 2 - 3 | 289 | 60 | |
BC137278.1 | - | 10 - 11 | 1082 | 60 | |
BC137280.1 | - | 10 - 11 | 1082 | 60 |