Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCH1
Entrez Gene ID
23007
Gene Name
phospholipase C eta 1
Gene Aliases
PLCL3
Chromosome Location
Chr.3: 155458603 - 155745723 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130960.1 NP_001124432.1 3 - 4 716 86
NM_001130961.1 NP_001124433.1 3 - 4 716 86
NM_014996.2 NP_055811.1 3 - 4 459 86
XM_005247238.1 XP_005247295.1 4 - 5 765 86
XM_005247239.1 XP_005247296.1 3 - 4 816 86
XM_011512560.2 XP_011510862.1 4 - 5 774 86
XM_011512561.2 XP_011510863.1 4 - 5 906 86
XM_011512562.2 XP_011510864.1 5 - 6 1523 86
XM_011512564.2 XP_011510866.1 4 - 5 765 86
XM_011512565.2 XP_011510867.1 3 - 4 1237 86
XM_011512566.2 XP_011510868.1 3 - 4 878 86
XM_011512567.1 XP_011510869.1 4 - 5 765 86
XM_017005923.1 XP_016861412.1 4 - 5 765 86
XM_017005924.1 XP_016861413.1 4 - 5 765 86
XM_017005925.1 XP_016861414.1 4 - 5 765 86
XM_017005926.1 XP_016861415.1 3 - 4 664 86
XM_017005927.1 XP_016861416.1 4 - 5 765 86
XM_017005928.1 XP_016861417.1 4 - 5 765 86
XM_017005929.1 XP_016861418.1 3 - 4 816 86
GenBank mRNA AK096620.1 - 3 - 4 816 86
AY691170.1 - 3 - 4 439 86
AY691171.1 - 3 - 4 439 86
BC113950.1 - 3 - 4 459 86

View Full Product Details