The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002646.3 | NP_002637.3 | 11 - 12 | 2226 | 56 |
XM_005245257.2 | XP_005245314.1 | 10 - 11 | 2291 | 56 | |
XM_005245258.3 | XP_005245315.1 | 10 - 11 | 2291 | 56 | |
XM_011509630.2 | XP_011507932.1 | 12 - 13 | 3085 | 56 | |
XM_011509631.2 | XP_011507933.1 | 11 - 12 | 2402 | 56 | |
XM_011509633.2 | XP_011507935.1 | 10 - 11 | 2291 | 56 | |
XM_017001473.1 | XP_016856962.1 | 11 - 12 | 3221 | 56 | |
XM_017001474.1 | XP_016856963.1 | 11 - 12 | 2424 | 56 | |
XM_017001475.1 | XP_016856964.1 | 9 - 10 | 1172 | 56 | |
GenBank mRNA | AB210006.1 | - | 9 - 10 | 1780 | 56 |
AK308153.1 | - | 12 - 13 | 2611 | 56 | |
BC132876.1 | - | 11 - 12 | 2198 | 56 | |
BC136645.1 | - | 11 - 12 | 2198 | 56 | |
BC144342.1 | - | 11 - 12 | 2198 | 56 | |
Y11312.1 | - | 11 - 12 | 2262 | 56 | |
Y13892.1 | - | 9 - 10 | 1763 | 56 |