Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSMD1
Entrez Gene ID
64478
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
PPP1R24
Chromosome Location
Chr.8: 2935353 - 4995035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033225.5 NP_150094.5 16 - 17 2801 90
XM_011534752.2 XP_011533054.1 16 - 17 3030 90
XM_017013731.1 XP_016869220.1 16 - 17 3030 90
GenBank mRNA AB209502.1 - 4 - 5 853 90
AF333704.2 - 17 - 18 2804 90
AK126857.1 - 6 - 7 1031 90
AY017307.1 - 17 - 18 2699 90

View Full Product Details