The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033225.5 | NP_150094.5 | 63 - 64 | 10206 | 108 |
XM_011534752.2 | XP_011533054.1 | 63 - 64 | 10435 | 108 | |
XM_011534753.2 | XP_011533055.1 | 46 - 47 | 7313 | 108 | |
XM_011534754.1 | XP_011533056.1 | 39 - 40 | 6049 | 108 | |
GenBank mRNA | AB209502.1 | - | 50 - 51 | 8069 | 108 |
AF333704.2 | - | 64 - 65 | 10212 | 108 | |
AK127129.1 | - | 14 - 15 | 2224 | 108 | |
AY017307.1 | - | 61 - 62 | 9576 | 108 |