The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244937.2 | NP_001231866.1 | 12 - 13 | 2941 | 68 |
NM_001318913.1 | NP_001305842.1 | 11 - 12 | 2770 | 68 | |
NM_002447.3 | NP_002438.2 | 13 - 14 | 3088 | 68 | |
NR_134919.1 | 13 - 14 | 3088 | 68 | ||
XM_005265170.4 | XP_005265227.2 | 13 - 14 | 3086 | 68 | |
XM_011533739.2 | XP_011532041.1 | 13 - 14 | 2966 | 68 | |
XM_011533740.2 | XP_011532042.1 | 12 - 13 | 2939 | 68 | |
XM_011533742.2 | XP_011532044.1 | 13 - 14 | 3086 | 68 | |
XM_011533743.2 | XP_011532045.1 | 13 - 14 | 3086 | 68 | |
XM_011533744.2 | XP_011532046.1 | 11 - 12 | 2768 | 68 | |
GenBank mRNA | AK303234.1 | - | 3 - 4 | 428 | 68 |
HQ908652.1 | - | 13 - 14 | 3060 | 68 | |
JN689382.1 | - | 3 - 4 | 885 | 68 | |
JN983806.1 | - | 11 - 12 | 2770 | 68 | |
X70040.1 | - | 13 - 14 | 3088 | 68 |