The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244937.2 | NP_001231866.1 | 4 - 5 | 1747 | 75 |
NM_001318913.1 | NP_001305842.1 | 2 - 3 | 1429 | 75 | |
NM_002447.3 | NP_002438.2 | 4 - 5 | 1747 | 75 | |
NR_134919.1 | 4 - 5 | 1747 | 75 | ||
XM_005265170.4 | XP_005265227.2 | 4 - 5 | 1742 | 75 | |
XM_011533739.2 | XP_011532041.1 | 4 - 5 | 1742 | 75 | |
XM_011533740.2 | XP_011532042.1 | 4 - 5 | 1742 | 75 | |
XM_011533741.2 | XP_011532043.1 | 4 - 5 | 1742 | 75 | |
XM_011533742.2 | XP_011532044.1 | 4 - 5 | 1742 | 75 | |
XM_011533743.2 | XP_011532045.1 | 4 - 5 | 1742 | 75 | |
XM_011533744.2 | XP_011532046.1 | 2 - 3 | 1424 | 75 | |
GenBank mRNA | EU826583.1 | - | 2 - 3 | 1401 | 75 |
EU826584.1 | - | 4 - 5 | 1719 | 75 | |
EU826585.1 | - | 4 - 5 | 1719 | 75 | |
HQ908652.1 | - | 4 - 5 | 1719 | 75 | |
JN983806.1 | - | 2 - 3 | 1429 | 75 | |
X70040.1 | - | 4 - 5 | 1747 | 75 |