The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145860.1 | NP_001139332.1 | 10 - 11 | 1571 | 73 |
NM_001145861.1 | NP_001139333.1 | 10 - 11 | 1511 | 73 | |
NM_015029.2 | NP_055844.2 | 10 - 11 | 1499 | 73 | |
XM_011516801.2 | XP_011515103.1 | 10 - 11 | 1571 | 73 | |
GenBank mRNA | AK291434.1 | - | 10 - 11 | 1568 | 73 |
BC011529.1 | - | 10 - 11 | 1499 | 73 | |
D31765.1 | - | 7 - 8 | 1110 | 73 |