Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3F
Entrez Gene ID
10621
Gene Name
RNA polymerase III subunit F
Gene Aliases
RPC39, RPC6
Chromosome Location
Chr.20: 18467389 - 18484648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282526.1 NP_001269455.1 8 - 9 1094 135
NM_006466.3 NP_006457.2 8 - 9 991 135
NR_104209.1 9 - 10 1049 135
XM_011529141.1 XP_011527443.1 9 - 10 1051 135
XM_017027596.1 XP_016883085.1 7 - 8 846 135
GenBank mRNA AK096684.1 - 9 - 10 1047 135
AK290892.1 - 8 - 9 962 135
AK314838.1 - 8 - 9 1093 135
BC012588.1 - 8 - 9 994 135
BC016761.1 - 8 - 9 1097 135

View Full Product Details