Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHC1
Entrez Gene ID
114327
Gene Name
EF-hand domain containing 1
Gene Aliases
EJM1, dJ304B14.2
Chromosome Location
Chr.6: 52420196 - 52495785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018100.3 NP_060570.2 1 - 2 272 59
NR_033327.1 1 - 2 272 59
GenBank mRNA AK001328.1 - 1 - 2 158 59
AK225543.1 - 1 - 2 166 59
AK295064.1 - 1 - 2 101 59
AL122084.1 - 1 - 2 159 59
AY608690.1 - 1 - 2 726 59
BC020210.1 - 1 - 2 148 59
DA906669.1 - 1 - 2 272 59
EU520261.1 - 1 - 2 157 59

View Full Product Details