The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172420.1 | NP_001165891.1 | 7 - 8 | 1258 | 78 |
NM_018100.3 | NP_060570.2 | 6 - 7 | 1346 | 78 | |
NR_033327.1 | 5 - 6 | 2603 | 78 | ||
GenBank mRNA | AK001328.1 | - | 6 - 7 | 1232 | 78 |
AK225543.1 | - | 5 - 6 | 2495 | 78 | |
AK295064.1 | - | 6 - 7 | 1089 | 78 | |
AK297632.1 | - | 7 - 8 | 1258 | 78 | |
AL122084.1 | - | 5 - 6 | 2489 | 78 | |
BC020210.1 | - | 6 - 7 | 1222 | 78 | |
EU520261.1 | - | 6 - 7 | 1231 | 78 |