The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144663.1 | NP_001138135.1 | 10 - 11 | 1415 | 170 |
NM_004063.3 | NP_004054.3 | 10 - 11 | 1414 | 170 | |
XM_011516790.1 | XP_011515092.1 | 10 - 11 | 1413 | 170 | |
GenBank mRNA | AK300638.1 | - | 2 - 3 | 293 | 170 |
AK301001.1 | - | 6 - 7 | 740 | 170 | |
AK312873.1 | - | 10 - 11 | 1415 | 170 | |
BC112013.1 | - | 10 - 11 | 1387 | 170 | |
BC113464.1 | - | 10 - 11 | 1375 | 170 | |
U07969.1 | - | 10 - 11 | 1376 | 170 | |
X83228.1 | - | 10 - 11 | 1381 | 170 |