The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018477.2 | NP_060947.1 | 7 - 8 | 702 | 67 |
XM_011536963.2 | XP_011535265.1 | 5 - 6 | 482 | 67 | |
GenBank mRNA | AF220190.1 | - | 7 - 8 | 679 | 67 |
AK000544.1 | - | 7 - 8 | 682 | 67 | |
AK022534.1 | - | 7 - 8 | 654 | 67 | |
AK315537.1 | - | 7 - 8 | 647 | 67 | |
BC005208.1 | - | 7 - 8 | 663 | 67 | |
BC011997.1 | - | 7 - 8 | 636 | 67 | |
BX161491.1 | - | 2 - 3 | 746 | 67 | |
BX248282.1 | - | 7 - 8 | 598 | 67 |